missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF6 Antibody (70B1413.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
264.00€
Specifications
| Antigen | ATF6 |
|---|---|
| Clone | 70B1413.1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 3 - 5 μg/mL, Chromatin Immunoprecipitation 1:10 - 1:500, Flow Cytometry 1 μg per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, CyTOF-ready |
| Applications | ChIP Assay |
Description
ATF6 Monoclonal specifically detects ATF6 in Human, Mouse, Rat, Porcine, Plant, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), CyTOF-ready, Knockout Validated.Specifications
| ATF6 | |
| 1 mg/mL | |
| ChIP Assay | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| TBS with 0.05% Sodium Azide | |
| activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
| ATF6 | |
| IgG1 κ | |
| Protein G purified | |
| This ATF6 antibody detects both the full length and the cleaved/active protein. |
| 70B1413.1 | |
| Western Blot 3 - 5 μg/mL, Chromatin Immunoprecipitation 1:10 - 1:500, Flow Cytometry 1 μg per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P18850 | |
| 22926 | |
| This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title