missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF6 Antibody (70B1413.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-40256SS
This item is not returnable.
View return policy
Description
ATF6 Monoclonal specifically detects ATF6 in Human, Mouse, Rat, Porcine, Plant, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), CyTOF-ready, Knockout Validated.
Specifications
| ATF6 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 3 - 5 μg/mL, Chromatin Immunoprecipitation 1:10 - 1:500, Flow Cytometry 1 μg per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50, Immunohistochemistry-Frozen 1:10 - 1:500, Immunoblotting, CyTOF-ready | |
| P18850 | |
| ATF6 | |
| This monoclonal antibody was made against a partial protein containing amino acids 1-273 of human ATF6. | |
| 0.025 mg | |
| Lipid and Metabolism, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR) | |
| 22926 | |
| Human | |
| Purified |
| ChIP Assay | |
| 70B1413.1 | |
| Unconjugated | |
| TBS with 0.05% Sodium Azide | |
| activating transcription factor 6, Activating transcription factor 6 alpha, atf6 a, ATF6A, ATF6-alpha, cAMP-dependent transcription factor ATF-6 alpha, cyclic AMP-dependent transcription factor ATF-6 alpha | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This ATF6 antibody detects both the full length and the cleaved/active protein. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction