missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL13B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 463.00€
Specifications
| Antigen | ARL13B |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631912
|
Novus Biologicals
NBP2-92454-0.02ml |
0.02 mL |
190.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637832
|
Novus Biologicals
NBP2-92454-0.1ml |
0.1 mL |
463.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARL13B Polyclonal antibody specifically detects ARL13B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ARL13B | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| GPCR, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 200894 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ADP-ribosylation factor-like 13B, ADP-ribosylation factor-like 2-like 1, ADP-ribosylation factor-like protein 13B, ADP-ribosylation factor-like protein 2-like 1, ARL2L1MGC120611, ARL2-like protein 1, DKFZp686E2075, DKFZp686L2472, DKFZp761H079, JBTS8DKFZp686M2074, MGC120612 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human ARL13B (NP_878899.1). MFSLMASCCGWFKRWREPVRKVTLLMVGLDNAGKTATAKGIQGEYPEDVAPTVGFSKINLRQGKFEVTIFDLGGGIRIRGIWKNYYAESYGVIFVVDSSDEERMEETKEAMSEMLRHPRISGKPILVLANKQDKEGALGEADVIECLSLEKLVNEHKCLCQIEPCSAISGYGKKIDKSIKKGLYWLLHVIARDFDALNERIQKETTEQRALEEQEKQERAERVRKLREERKQNEQEQAEL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title