missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL13B Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92454-0.1ml
This item is not returnable.
View return policy
Description
ARL13B Polyclonal antibody specifically detects ARL13B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ARL13B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| ADP-ribosylation factor-like 13B, ADP-ribosylation factor-like 2-like 1, ADP-ribosylation factor-like protein 13B, ADP-ribosylation factor-like protein 2-like 1, ARL2L1MGC120611, ARL2-like protein 1, DKFZp686E2075, DKFZp686L2472, DKFZp761H079, JBTS8DKFZp686M2074, MGC120612 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human ARL13B (NP_878899.1). MFSLMASCCGWFKRWREPVRKVTLLMVGLDNAGKTATAKGIQGEYPEDVAPTVGFSKINLRQGKFEVTIFDLGGGIRIRGIWKNYYAESYGVIFVVDSSDEERMEETKEAMSEMLRHPRISGKPILVLANKQDKEGALGEADVIECLSLEKLVNEHKCLCQIEPCSAISGYGKKIDKSIKKGLYWLLHVIARDFDALNERIQKETTEQRALEEQEKQERAERVRKLREERKQNEQEQAEL | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 200894 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction