missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argonaute 3 Antibody (4F7), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Argonaute 3 |
|---|---|
| Clone | 4F7 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043158
|
Novus Biologicals
NBP3-26519-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043629
|
Novus Biologicals
NBP3-26519-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Argonaute 3 Monoclonal antibody specifically detects Argonaute 3 in Human samples. It is validated for ELISASpecifications
| Argonaute 3 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 192669 | |
| IgG | |
| Affinity purified |
| 4F7 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| AGO3argonaute 3, argonaute3, eIF2C 3, eIF-2C 3, Eukaryotic translation initiation factor 2C 3, eukaryotic translation initiation factor 2C, 3, FLJ12765, hAgo3, MGC86946, protein argonaute-3 | |
| A synthesized peptide derived from Human Argonaute 3 [UniProt Q9H9G7] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title