missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argonaute 3 Antibody (4F7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26519-50ul
This item is not returnable.
View return policy
Description
Argonaute 3 Monoclonal antibody specifically detects Argonaute 3 in Human samples. It is validated for ELISA
Specifications
| Argonaute 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 192669 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 4F7 | |
| ELISA | |
| AGO3argonaute 3, argonaute3, eIF2C 3, eIF-2C 3, Eukaryotic translation initiation factor 2C 3, eukaryotic translation initiation factor 2C, 3, FLJ12765, hAgo3, MGC86946, protein argonaute-3 | |
| A synthesized peptide derived from Human Argonaute 3 [UniProt Q9H9G7] | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction