missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€ - 544.00€
Specifications
| Antigen | ARF6 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18693559
|
Novus Biologicals
NBP2-41263-0.025mg |
0.025 mg |
189.00€
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18124259
|
Novus Biologicals
NBP2-41263 |
0.1 mg |
544.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARF6 Polyclonal specifically detects ARF6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| ARF6 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| ADP-ribosylation factor 6, DKFZp564M0264 | |
| ARF6 | |
| IgG | |
| Affinity Purified | |
| 18 kDa |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 382 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human ARF6. The immunogen is located within the last 50 amino acids of ARF6. Amino Acid Squence: PHEIQEKLGLTRIRDRNWY | |
| Primary | |
| ARF6 antibody is human, mouse and rat reactive. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title