missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41263
This item is not returnable.
View return policy
Description
ARF6 Polyclonal specifically detects ARF6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| ARF6 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| ARF6 | |
| Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human ARF6. The immunogen is located within the last 50 amino acids of ARF6. Amino Acid Squence: PHEIQEKLGLTRIRDRNWY | |
| Affinity Purified | |
| RUO | |
| Primary | |
| ARF6 antibody is human, mouse and rat reactive. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml | |
| ADP-ribosylation factor 6, DKFZp564M0264 | |
| Rabbit | |
| 18 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication, Signal Transduction | |
| 382 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction