missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29906-0.025mg
This item is not returnable.
View return policy
Description
ARF1 Polyclonal specifically detects ARF1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| ARF1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1.0 mg/mL BSA and 30% Glycerol with 0.05% Sodium Azide | |
| ADP-ribosylation factor 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 375 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:10 - 1:100 | |
| P84077, P84075 | |
| ARF1 | |
| Synthetic peptide DWLSNQLRNQK | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Canine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction