missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00€
Specifications
| Antigen | ARF1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:10 - 1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
ARF1 Polyclonal specifically detects ARF1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| ARF1 | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10 - 1:100, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:10 - 1:100 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Canine | |
| P84077, P84075 | |
| 375 | |
| Synthetic peptide DWLSNQLRNQK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, 1.0 mg/mL BSA and 30% Glycerol with 0.05% Sodium Azide | |
| ADP-ribosylation factor 1 | |
| ARF1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title