missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-5 Antibody (SR1459), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00€ - 449.00€
Specifications
| Antigen | Aquaporin-5 |
|---|---|
| Clone | SR1459 |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692625
|
Novus Biologicals
NBP3-21684-100ul |
100 μg |
449.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645964
|
Novus Biologicals
NBP3-21684-50ul |
50 μg |
349.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aquaporin-5 Monoclonal antibody specifically detects Aquaporin-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| Aquaporin-5 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AQP-5, aquaporin 5, aquaporin-5 | |
| A synthesized peptide derived from human Aquaporin-5 (Uniprot #: P55064) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1459 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 362 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel