missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-5 Antibody (SR1459), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21684-100ul
This item is not returnable.
View return policy
Description
Aquaporin-5 Monoclonal antibody specifically detects Aquaporin-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| Aquaporin-5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1459 | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| AQP-5, aquaporin 5, aquaporin-5 | |
| A synthesized peptide derived from human Aquaporin-5 (Uniprot #: P55064) | |
| 100 μg | |
| Cancer, Signal Transduction | |
| 362 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction