missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Aprataxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00€ - 508.00€
Specifications
| Antigen | Aprataxin |
|---|---|
| Dilution | Western Blot : 1:400, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067125
|
Novus Biologicals™
NBP3-30957-25ug |
25 μg |
213.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067225
|
Novus Biologicals™
NBP3-30957-100ug |
100 μg |
508.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aprataxin Polyclonal antibody specifically detects Aprataxin in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Aprataxin | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, Tumor Suppressors | |
| Phosphate buffered saline | |
| 54840 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1:400, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AOA1EAOH, AOAMGC1072, aprataxin, ataxia 1, early onset with hypoalbuminemia, AXA1, EC 3.-, EOAHA, FHA-HIT, FLJ20157, Forkhead-associated domain histidine triad-like protein | |
| Synthetic peptide derived from the human Aprataxin protein. | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title