missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Aprataxin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30957-25ug
This item is not returnable.
View return policy
Description
Aprataxin Polyclonal antibody specifically detects Aprataxin in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| Aprataxin | |
| Polyclonal | |
| Western Blot : 1:400, ELISA | |
| AOA1EAOH, AOAMGC1072, aprataxin, ataxia 1, early onset with hypoalbuminemia, AXA1, EC 3.-, EOAHA, FHA-HIT, FLJ20157, Forkhead-associated domain histidine triad-like protein | |
| Synthetic peptide derived from the human Aprataxin protein. | |
| 25 μg | |
| Cancer, DNA Repair, Tumor Suppressors | |
| 54840 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction