Learn More
TNF Mouse anti-Human, FITC, Clone: MAB11, BD
Mouse Monoclonal Antibody
213.00€ - 320.00€
Specifications
| Antigen | TNF |
|---|---|
| Clone | MAB11 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | FITC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15837748
|
BD Biosciences
554512 |
0.1 mg |
320.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15836898
|
BD Biosciences
552889 |
50 Tests |
213.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MAb11 antibody reacts with human tumor necrosis factor (TNF, formerly known as TNF-α) protein. TNF is an efficient paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The use of the MAb11 antibody has been reported to cross-react with TNF of rhesus monkey.
Host Species: Mouse
Clone: MAb11
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Recombinant Human TNF
Flow Cytometry, Intracellular Staining
Specifications
| TNF | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Recombinant Human TNF | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| MAB11 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.