Learn More
TNF Mouse anti-Human, FITC, Clone: MAB11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554512
Description
The MAb11 antibody reacts with human tumor necrosis factor (TNF, formerly known as TNF-α) protein. TNF is an efficient paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The use of the MAb11 antibody has been reported to cross-react with TNF of rhesus monkey.
Host Species: Mouse
Clone: MAb11
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Recombinant Human TNF
Flow Cytometry, Intracellular Staining
Specifications
| TNF | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human TNF | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| MAB11 | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.