Learn More
PKA[C] Mouse, Unlabeled, Clone: 5B, BD
Mouse Monoclonal Antibody
211.00€ - 359.00€
Specifications
| Antigen | PKA[C] |
|---|---|
| Clone | 5B |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15875739
|
BD Biosciences
610980 |
50 μg |
211.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15885739
|
BD Biosciences
610981 |
150 μg |
359.00€
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
cAMP-dependent Protein Kinase (PKA) is composed of two distinct subunits: catalytic (C) and regulatory (R). Four regulatory subunits have been identified: RIα, RIβ, RIIα, and RIIβ. These subunits define type I and II cAMP-dependent protein kinases. Following binding of cAMP, the regulatory subunits dissociate from the catalytic subunits, rendering the enzyme active. Type I and type II holoenzymes have three potential C subunits (Cα, Cβ, or Cγ). Type II PKA can be distinguished by autophosphorylation of the R-subunits, while type I PKA binds Mg/ATP with high affinity. The levels of expression of the different subunits vary according to cell and tissue type.
Immunofluorescence, Western Blotting
Specifications
| PKA[C] | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2b | |
| Affinity Purified |
| 5B | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| Human PKA[Cα] subunit aa. 18-347 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.