Learn More
Anti-Human CD71 (OKT9), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65235-1-IG-500UG
This item is not returnable.
View return policy
Description
The CD71 antibody from Proteintech is a mouse monoclonal antibody generated with human t-cell acute lymphoblastic leukemia cells of human CD71. This antibody recognizes human antigen. The CD71 antibody has been validated for the following applications: FC analysis.Specifications
| CD71 | |
| Monoclonal | |
| 0.5 mg/mL | |
| CD71; TFR; TFR1; TRFR | |
| Human T-cell acute lymphoblastic leukemia cells | |
| 500 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| OKT9 | |
| Unconjugated | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 7037 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |