missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human CD71 (OKT9), Proteintech

Mouse Monoclonal Antibody

55.00€ - 168.00€

Specifications

Antigen CD71
Clone OKT9
Concentration 0.5 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234005
View Documents
Proteintech
65235-1-IG-100UG
100 μg
ONLINE EXCLUSIVE PRICE
58.00€
55.00€ / 100µg
Save 3.00€
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30235122
View Documents
Proteintech
65235-1-IG-500UG
500 μg
ONLINE EXCLUSIVE PRICE
179.00€
168.00€ / 500µg
Save 11.00€
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The CD71 antibody from Proteintech is a mouse monoclonal antibody generated with human t-cell acute lymphoblastic leukemia cells of human CD71. This antibody recognizes human antigen. The CD71 antibody has been validated for the following applications: FC analysis.
Specifications

Specifications

CD71
0.5 mg/mL
Monoclonal
Liquid
RUO
7037
Human T-cell acute lymphoblastic leukemia cells
Primary
Store at 2-8°C. Stable for one year after shipment.
OKT9
Flow Cytometry
Unconjugated
Mouse
Human
CD71; TFR; TFR1; TRFR
IgG1 κ
Affinity purification
Flow Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.