Learn More
Anti-Human CD71 (OKT9), Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 65235-1-IG-100UG
 View more versions of this product
Product Code. 30234005
  • ONLINE EXCLUSIVE PRICE
    58.00€
    55.00€ / 100µg
    Save 3.00€
    5% Off
Description
The CD71 antibody from Proteintech is a mouse monoclonal antibody generated with human t-cell acute lymphoblastic leukemia cells of human CD71. This antibody recognizes human antigen. The CD71 antibody has been validated for the following applications: FC analysis.
Specifications
Specifications
CD71
Monoclonal
0.5 mg/mL
CD71; TFR; TFR1; TRFR
Human T-cell acute lymphoblastic leukemia cells
100 μg
Primary
Human
Flow Antibody
IgG1 κ
Flow Cytometry
OKT9
Unconjugated
Mouse
Affinity purification
RUO
7037
Store at 2-8°C. Stable for one year after shipment.