missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD11a (TS2/4), Proteintech
Mouse Monoclonal Antibody
50.00€ - 152.00€
Specifications
| Antigen | CD11a |
|---|---|
| Clone | TS2/4 |
| Concentration | 500 μg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234734
|
Proteintech
65194-1-IG-500UG |
500 μg |
161.00€ 152.00€ / 500µg Save 9.00€ 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30238031
|
Proteintech
65194-1-IG-100UG |
100 μg |
52.00€ 50.00€ / 100µg Save 2.00€ 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD11a antibody from Proteintech is a mouse monoclonal antibody generated with hla-dr ctl line of human CD11a. This antibody recognizes human antigen. The CD11a antibody has been validated for the following applications: FC analysis.Specifications
| CD11a | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 3683 | |
| HLA-DR CTL line | |
| Primary | |
| Store at 2-8°C. Stable for one year after shipment. |
| TS2/4 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD11A; LFA-1; LFA1A | |
| IgG1 κ | |
| Protein G purification | |
| Flow Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title