missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Human CD11a (TS2/4), Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 65194-1-IG-100UG
This item is not returnable.
View return policy
Description
The CD11a antibody from Proteintech is a mouse monoclonal antibody generated with hla-dr ctl line of human CD11a. This antibody recognizes human antigen. The CD11a antibody has been validated for the following applications: FC analysis.Specifications
| CD11a | |
| Monoclonal | |
| 500 μg/mL | |
| CD11A; LFA-1; LFA1A | |
| HLA-DR CTL line | |
| 100 μg | |
| Primary | |
| Human | |
| Flow Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| TS2/4 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3683 | |
| Store at 2-8°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction