missing translation for 'onlineSavingsMsg'
Learn More

CD11b/Mac-1 Mouse anti-Human, APC, Clone: ICRF44, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 550019

 View more versions of this product

Product Code. 15856168

  • ONLINE EXCLUSIVE PRICE
    377.00€
    376.68€ / Each
    Save 0.32€
    0.09% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reacts with the 165kDa adhesion glycoprotein (Mac-1, integrin αM subunit) which associates with the 95kDa integrin β2 (CD18) to form the CD11b/CD18 complex. CD11b/Mac-1 is expressed on activated lymphocytes, monocytes, granulocytes, and a subset of NK cells. CD11b functions in cell-cell and cell-substrate interactions and as a receptor for iC3b, CD54 (ICAM-1), CD102 (ICAM-2) and CD50 (ICAM-3). This antibody significantly inhibits PMN aggregation in response to fMLP. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: ICRF44
Isotype: IgG1, κ
Cross Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD11b
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human monocytes
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
ICRF44
MAC-1A; Mac-1; ITGAM; Integrin alpha M; CR3A; CR-3 alpha; Mo1; SLEB6
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.