missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase P2/XPNPEP2 Rabbit anti-Human, Clone: 18, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00€ - 552.00€
Specifications
| Antigen | Aminopeptidase P2/XPNPEP2 |
|---|---|
| Clone | 18 |
| Dilution | Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263396
|
Novus Biologicals
NBP2-90014-50UL |
50 μg |
367.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18238446
|
Novus Biologicals
NBP2-90014-100UL |
100 μg |
552.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase P2/XPNPEP2 Monoclonal specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for Sandwich ELISA Detection.Specifications
| Aminopeptidase P2/XPNPEP2 | |
| Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 7512 | |
| IgG | |
| Protein A purified |
| 18 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound | |
| Recombinant Human Aminopeptidase P2/XPNPEP2 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title