missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase P2/XPNPEP2 Rabbit anti-Human, Clone: 18, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP2-90014-100UL

 View more versions of this product

Product Code. 18238446

  • 552.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Aminopeptidase P2/XPNPEP2 Monoclonal specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for Sandwich ELISA Detection.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aminopeptidase P2/XPNPEP2
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Protein A purified
RUO
7512
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
ELISA
18
Sandwich ELISA Detection 1:1000-1:10000
Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
Recombinant Human Aminopeptidase P2/XPNPEP2 Protein
100 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.