missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody (AP1B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
214.00€
Specifications
| Antigen | Alkaline Phosphatase, Intestinal |
|---|---|
| Clone | AP1B9 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
Description
Alkaline Phosphatase, Intestinal Monoclonal specifically detects Alkaline Phosphatase, Intestinal in Human, Bovine samples. It is validated for Immunohistochemistry.Specifications
| Alkaline Phosphatase, Intestinal | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Embryonic Stem Cell Markers, Stem Cell Markers | |
| alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
| ALPI | |
| IgG1 | |
| Protein A purified | |
| calf intestinal Alkaline Phosphatase (AP1B9). |
| AP1B9 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 248 | |
| Calf Alkaline phosphatase | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title