missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody (AP1B9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2637-0.025mg
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Intestinal Monoclonal specifically detects Alkaline Phosphatase, Intestinal in Human, Bovine samples. It is validated for Immunohistochemistry.
Specifications
| Alkaline Phosphatase, Intestinal | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:10 - 1:500 | |
| ALPI | |
| Calf Alkaline phosphatase | |
| 0.025 mg | |
| Embryonic Stem Cell Markers, Stem Cell Markers | |
| 248 | |
| Human | |
| Purified |
| Immunohistochemistry | |
| AP1B9 | |
| Unconjugated | |
| alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| calf intestinal Alkaline Phosphatase (AP1B9). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction