Learn More
Alix Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 450.00€
Specifications
| Antigen | Alix |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006405
|
Proteintech
CL488-12422-20UL |
20 μL |
140.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18046715
|
Proteintech
CL488-12422-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Alix antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Alix. This antibody recognizes human, mouse, rat antigen. The Alix antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| Alix | |
| Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| AIP1, ALG 2 interacting protein 1, Alix, DRIP4, HP95, KIAA1375, PDCD6 interacting protein, PDCD6IP | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 10015 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.