Learn More
Alix Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-12422-20UL
 View more versions of this product
Product Code. 18006405
  • 140.00€ / 20µL
Description
The Alix antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Alix. This antibody recognizes human, mouse, rat antigen. The Alix antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
Alix
1000 μg/mL
AIP1, ALG 2 interacting protein 1, Alix, DRIP4, HP95, KIAA1375, PDCD6 interacting protein, PDCD6IP
20 μL
10015
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa