missing translation for 'onlineSavingsMsg'
Learn More

ALDH1A2 Antibody (SR2014), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00€ - 449.00€

Specifications

Antigen ALDH1A2
Clone SR2014
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18681145
View Documents
Novus Biologicals
NBP3-21945-100ul
100 μg
449.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18653045
View Documents
Novus Biologicals
NBP3-21945-50ul
50 μg
338.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ALDH1A2 Monoclonal antibody specifically detects ALDH1A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

ALDH1A2
Western Blot 1:500-1:2000
Monoclonal
Purified
RUO
Human, Mouse, Rat
aldehyde dehydrogenase 1 family, member A2, Aldehyde dehydrogenase family 1 member A2, EC 1.2.1, EC 1.2.1.36, RALDH 2, RALDH(II), RalDH2, RALDH2MGC26444, RALDH2-T, retinal dehydrogenase 2, Retinaldehyde-specific dehydrogenase type 2
A synthesized peptide derived from human ALDH1A2 (Uniprot #: O94788)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2014
Western Blot
Unconjugated
Rabbit
Cell Biology, Neuroscience
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
8854
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.