missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A2 Antibody (SR2014), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21945-50ul
This item is not returnable.
View return policy
Description
ALDH1A2 Monoclonal antibody specifically detects ALDH1A2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ALDH1A2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR2014 | |
| Western Blot 1:500-1:2000 | |
| aldehyde dehydrogenase 1 family, member A2, Aldehyde dehydrogenase family 1 member A2, EC 1.2.1, EC 1.2.1.36, RALDH 2, RALDH(II), RalDH2, RALDH2MGC26444, RALDH2-T, retinal dehydrogenase 2, Retinaldehyde-specific dehydrogenase type 2 | |
| A synthesized peptide derived from human ALDH1A2 (Uniprot #: O94788) | |
| 50 μg | |
| Cell Biology, Neuroscience | |
| 8854 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction