missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF-1/Iba1 Antibody (SR1261), Novus Biologicals™
Rabbit Monoclonal Antibody
349.00€ - 472.00€
Specifications
| Antigen | AIF-1/Iba1 |
|---|---|
| Clone | SR1261 |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666124
|
Novus Biologicals
NBP3-21860-100ul |
100 μg |
472.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620754
|
Novus Biologicals
NBP3-21860-50ul |
50 μg |
349.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| AIF-1/Iba1 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
| A synthesized peptide derived from human AIF-1/Iba1 (Uniprot #: P55008) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1261 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 199 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title