missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF-1/Iba1 Antibody (SR1261), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21860-100ul
This item is not returnable.
View return policy
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| AIF-1/Iba1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1261 | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
| A synthesized peptide derived from human AIF-1/Iba1 (Uniprot #: P55008) | |
| 100 μg | |
| Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
| 199 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur