missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADTB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Specifications
| Antigen | ADTB1 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunoprecipitation 0.1-0.5 uL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614126
|
Novus Biologicals
NBP2-99079-100ul |
100 μg |
326.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18608405
|
Novus Biologicals
NBP2-99079-50ul |
50 μg |
216.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADTB1 Polyclonal antibody specifically detects ADTB1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| ADTB1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Membrane Vesicle Markers | |
| 0.2 μm filtered solution in PBS | |
| 162 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.1-0.5 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Adapter-Related Protein Complex 1 Subunit Beta-1, Adaptor Protein Complex AP-1 Subunit Beta-1, Adaptor-Related Protein Complex 1, Beta 1 Subunit, AP-1 Complex Subunit Beta-1, AP105A, AP1B1, BAM22, beta1-adaptin, Beta-1-adaptin, Beta-Adaptin 1, beta-prime-adaptin, CLAPB2, Clathrin Assembly Protein Complex 1 Beta Large Chain, Golgi Adaptor HA1/AP1 Adaptin Beta Subunit, Plasma Membrane Adaptor HA2/AP2 Adaptor Beta Subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ADTB1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title