missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADTB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99079-100ul
This item is not returnable.
View return policy
Description
ADTB1 Polyclonal antibody specifically detects ADTB1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| ADTB1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunoprecipitation 0.1-0.5 uL/mg of lysate | |
| Adapter-Related Protein Complex 1 Subunit Beta-1, Adaptor Protein Complex AP-1 Subunit Beta-1, Adaptor-Related Protein Complex 1, Beta 1 Subunit, AP-1 Complex Subunit Beta-1, AP105A, AP1B1, BAM22, beta1-adaptin, Beta-1-adaptin, Beta-Adaptin 1, beta-prime-adaptin, CLAPB2, Clathrin Assembly Protein Complex 1 Beta Large Chain, Golgi Adaptor HA1/AP1 Adaptin Beta Subunit, Plasma Membrane Adaptor HA2/AP2 Adaptor Beta Subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ADTB1. | |
| 100 μg | |
| Membrane Vesicle Markers | |
| 162 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction