missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS1 Antibody (1G5D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 552.00€
Specifications
| Antigen | ADAMTS1 |
|---|---|
| Clone | 1G5D5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18299504
|
Novus Biologicals
NBP2-61808 |
0.1 mL |
552.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683927
|
Novus Biologicals
NBP2-61808-0.025ml |
0.025 mL |
203.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAMTS1 Monoclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| ADAMTS1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH-1 | |
| ADAMTS1 | |
| IgG1 | |
| Protein G purified |
| 1G5D5 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| Cell Cycle and Replication | |
| 9510 | |
| Purified recombinant fragment of human ADAMTS1 (AA: 858-960) expressed in E. Coli. | |
| Primary | |
| 147 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title