missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS1 Antibody (1G5D5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61808
This item is not returnable.
View return policy
Description
ADAMTS1 Monoclonal specifically detects ADAMTS1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ADAMTS1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 1, ADAM metallopeptidase with thrombospondin type 1 motif, 1, ADAM-TS 1, ADAM-TS1, ADAMTS-1, C3-C5, METH-1 | |
| Mouse | |
| 147 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 1G5D5 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ADAMTS1 | |
| Purified recombinant fragment of human ADAMTS1 (AA: 858-960) expressed in E. Coli. | |
| Protein G purified | |
| Cell Cycle and Replication | |
| 9510 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction