missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein Antibody (2H2), Novus Biologicals™
Mouse Monoclonal Antibody
185.00€ - 609.00€
Specifications
| Antigen | Acrolein |
|---|---|
| Clone | 2H2 |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18230713
|
Novus Biologicals
NBP2-59358 |
100 μg |
609.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691737
|
Novus Biologicals
NBP2-59358-25ug |
25 μg |
185.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acrolein Monoclonal specifically detects Acrolein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Acrolein | |
| Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins. |
| 2H2 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title