missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein Antibody (2H2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59358-25ug
This item is not returnable.
View return policy
Description
Acrolein Monoclonal specifically detects Acrolein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Acrolein | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Specific for Acrolein modified proteins. Does not detect free acrolein. Does not cross-react with Crotonaldehyde, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins. | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 2H2 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Acrylic amide; Ethylene Carboxamide 2-Propenamide; 2-Propen-1-one; 2-propenal; Acraldehyde; Acrolein; Acrylic aldehyde | |
| Synthetic Acrolein modified Keyhole Limpet Hemocyanin (KLH). | |
| 25 μg | |
| Primary | |
| Store at -20°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction