missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG5 Antibody (1B5E10), Novus Biologicals™
Mouse Monoclonal Antibody
226.00€ - 449.00€
Specifications
| Antigen | ABCG5 |
|---|---|
| Clone | 1B5E10 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:2000, Immunohistochemistry-Paraffin 1:200 - 1:2000, ICC/IF (Negative) |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18107851
|
Novus Biologicals
NBP2-23488SS |
0.025 mL |
226.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18177141
|
Novus Biologicals
NBP2-23488 |
0.1 mL | |||||||
Description
ABCG5 Monoclonal specifically detects ABCG5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).Specifications
| ABCG5 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| ATP-binding cassette sub-family G member 5, ATP-binding cassette, sub-family G (WHITE), member 5, ATP-binding cassette, subfamily G, member 5, sterolin 1, sterolin-1, STSL | |
| ABCG5 | |
| IgG1 | |
| Ammonium sulfate precipitation |
| 1B5E10 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:2000, Immunohistochemistry-Paraffin 1:200 - 1:2000, ICC/IF (Negative) | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Q9H222 | |
| 64240 | |
| Partial recombinant human ABCG5 (between residues 300-500) expressed in E. coli [UniProt Q9H222]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title