missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCG5 Antibody (1B5E10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23488
This item is not returnable.
View return policy
Description
ABCG5 Monoclonal specifically detects ABCG5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, ICC/IF (Negative).
Specifications
| ABCG5 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:2000, Immunohistochemistry-Paraffin 1:200 - 1:2000, ICC/IF (Negative) | |
| ATP-binding cassette sub-family G member 5, ATP-binding cassette, sub-family G (WHITE), member 5, ATP-binding cassette, subfamily G, member 5, sterolin 1, sterolin-1, STSL | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| 64240 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1B5E10 | |
| Unconjugated | |
| Q9H222 | |
| ABCG5 | |
| Partial recombinant human ABCG5 (between residues 300-500) expressed in E. coli [UniProt Q9H222]. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction