missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAV5 VP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93577SS
This item is not returnable.
View return policy
Description
AAV5 VP3 Polyclonal specifically detects AAV5 VP3 in Non-species specific, Virus samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| AAV5 VP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Adeno-Associated Virus, Adeno-Associated Virus (AAV) | |
| Synthetic peptide within residues 525-550 of 80 kDa capsid VP3 protein of AAV5. | |
| 0.025 mg | |
| Primary | |
| Virus, Non-species specific | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry | |
| Q9YIJ1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Adeno-Associated Virus (AAV) | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction