missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AAV5 VP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
226.00€ - 488.00€
Specifications
| Antigen | AAV5 VP3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18290334
|
Novus Biologicals
NB100-93577SS |
0.025 mg |
226.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18726063
|
Novus Biologicals
NB100-93577 |
0.1 mg |
488.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AAV5 VP3 Polyclonal specifically detects AAV5 VP3 in Non-species specific, Virus samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| AAV5 VP3 | |
| Western Blot 1:500-1:1000, Immunohistochemistry | |
| Polyclonal | |
| Rabbit | |
| Virus, Non-species specific | |
| Q9YIJ1 | |
| Synthetic peptide within residues 525-550 of 80 kDa capsid VP3 protein of AAV5. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Adeno-Associated Virus, Adeno-Associated Virus (AAV) | |
| IgG | |
| Affinity Purified | |
| Adeno-Associated Virus (AAV) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title