Learn More
8 Regulated Phthalates and BPA in Medium Density Polyethylene, SPEX CertiPrep™
Shop All SPEX Certiprep Products
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
missing translation for 'unitSize'
5g
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Diisodecyl phthalate | 26761-40-0 | 30,000μg/g |
| Diisononyl phthalate | 28553-12-0 | 30,000μg/g |
| Bis(2-Ethylhexyl)phthalate | 117-81-7 | 3000μg/g |
| Bisphenol A | 80-05-7 | 3000μg/g |
| Butylbenzyl phthalate | 85-68-7 | 3000μg/g |
| Diethyl phthalate | 84-66-2 | 3000μg/g |
| Dimethyl phthalate | 131-11-3 | 3000μg/g |
| Di-n-butyl phthalate | 84-74-2 | 3000μg/g |
| Di-n-octyl phthalate | 117-84-0 | 3000μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | 8 Regulated Phthalates and BPA in Medium Density PE |
| Packaging | 2mL Amber Prescored Glass Ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| Technique | AA, ICP |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Quantity | 5 g |
Safety and Handling
Product Identifier
- Phthalates in Polyethylene Standard w/BPA
- Danger
- Carcinogenicity Category 2
- Reproductive toxicity Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : di-n-octyl phthalate, bisphenol A, BBP,dibutyl phthalate, bis(2-ethylhexyl) phthalate
Product intended for laboratory use only