Learn More
4-Amino-5-[(3,4,5-trimethoxyphenyl)methyl]-2(1H)-pyrimidinone, TRC
Product Code. 30135330
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135330 100 mg 100mg
17922655 10 mg 10mg
17922375 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135330 Supplier Toronto Research Chemicals Supplier No. TRCA630890100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 60729-91-1
Molecular Formula C14 H17 N3 O4
Molecular Weight (g/mol) 291.3
Synonym 4-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-2-ol, Trimethoprim Imp. E (EP)
IUPAC Name 4-amino-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidin-2-ol
SMILES COc1cc(Cc2cnc(O)nc2N)cc(OC)c1OC
Specifications
Analyte or Component Names 4-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-2-ol
Shipping Condition Room Temperature
InChI Formula InChI=1S/C14H17N3O4/c1-19-10-5-8(6-11(20-2)12(10)21-3)4-9-7-16-14(18)17-13(9)15/h5-7H,4H2,1-3H3,(H3,15,16,17,18)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 291.1219
Percent Purity >95%
Chemical Name or Material 4-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-2-ol
Grade HPLC