CAS RN 60729-91-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-2-ol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-Amino-5-[(3,4,5-trimethoxyphenyl)methyl]-2(1H)-pyrimidinone, TRC
CAS: 60729-91-1 Molecular Formula: C14 H17 N3 O4 Molecular Weight (g/mol): 291.3 Synonym: 4-Amino-5-(3,4,5-trimethoxybenzyl)pyrimidin-2-ol,Trimethoprim Imp. E (EP) IUPAC Name: 4-amino-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidin-2-ol SMILES: COc1cc(Cc2cnc(O)nc2N)cc(OC)c1OC