Learn More
3'-(Trifluoromethyl)acetophenone Oxime, TRC
Product Code. 30452558
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452558 1 g -
30456327 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30452558 Supplier Toronto Research Chemicals Supplier No. TRCT7905701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99705-50-7
Molecular Formula C9H8F3NO
Molecular Weight (g/mol) 203.16
Synonym m-(Trifluoromethyl)acetophenone Oxime, 1-[3-(Trifluoromethyl)phenyl]ethanone Oxime
IUPAC Name (NE)-N-[1-[3-(trifluoromethyl)phenyl]ethylidene]hydroxylamine
SMILES FC(F)(F)C1=CC=CC(/C(C)=N/O)=C1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H8F3NO/c1-6(13-14)7-3-2-4-8(5-7)9(10,11)12/h2-5,14H,1H3/b13-6+
Formula Weight 203.0558
Chemical Name or Material 3'-(Trifluoromethyl)acetophenone Oxime