CAS RN 99705-50-7
CAS RN 99705-50-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3'-(Trifluoromethyl)acetophenone Oxime, TRC
CAS: 99705-50-7 Molecular Formula: C9H8F3NO Molecular Weight (g/mol): 203.16 Synonym: m-(Trifluoromethyl)acetophenone Oxime,1-[3-(Trifluoromethyl)phenyl]ethanone Oxime IUPAC Name: (NE)-N-[1-[3-(trifluoromethyl)phenyl]ethylidene]hydroxylamine SMILES: FC(F)(F)C1=CC=CC(/C(C)=N/O)=C1