missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15455457
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15455457

Brand: Thermo Scientific Alfa Aesar H32703.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-92-3
Molecular Formula C9H6ClF3O2
Molecular Weight (g/mol) 238.59
MDL Number MFCD09025410
InChI Key IHMFMHOCMZSKNI-UHFFFAOYSA-N
Synonym 3-methoxy-5-trifluoromethyl benzoyl chloride, 5-methoxy-3-trifluoromethyl benzoyl chloride, benzoyl chloride, 3-methoxy-5-trifluoromethyl
PubChem CID 46737576
IUPAC Name 3-methoxy-5-(trifluoromethyl)benzoyl chloride
SMILES COC1=CC(=CC(=C1)C(F)(F)F)C(=O)Cl

Specifications

Refractive Index 1.4915
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 238.59
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-(trifluoromethyl)benzoyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.