accessibility menu, dialog, popup

UserName

CAS RN 916420-92-3

CH3OFFFOCl

CAS RN 916420-92-3

IUPAC Name: 3-methoxy-5-(trifluoromethyl)benzoyl chloride
Synonyms: 3-methoxy-5-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: IHMFMHOCMZSKNI-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(=C1)C(F)(F)F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results